siRNA Details
Gene Symbol: Olfr809 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146324.1 | NP_666436.1 | 1 | 935 | ||
GenBank mRNA | BC120625.1 | - | 1 | 979 | 40129548 | |
BC137816.1 | - | 1 | 979 | 8860835 |
Gene Symbol: Olfr813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207147.1 | NP_997030.1 | 1 | 890 | ||
GenBank mRNA | BC127959.1 | - | 1 | 894 | 40134789 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001001386, NM_001001363.
Gene Ontology Categories:
