siRNA Details
Gene Symbol: Olfr1284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146381.1 | NP_666493.1 | 1 | 373 | ||
GenBank mRNA | BC116802.1 | - | 1 | 408 | 40126121 | |
BC116804.1 | - | 1 | 408 | 40126123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
