siRNA Details
Gene Symbol: Olfr208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146384.1 | NP_666496.1 | 1 | 815 |
Gene Symbol: Olfr207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011792.1 | NP_001011792.1 | 1 | 815 |
Gene Symbol: Olfr209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207551.2 | NP_997434.2 | 1 | 815 |

More Information
Other Species Targeted

Gene Ontology Categories:
