siRNA Details
Gene Symbol: Olfr1392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146470.2 | NP_666681.1 | 1 | 460 | ||
GenBank mRNA | BC120614.1 | - | 1 | 456 | 40129537 | |
BC120616.1 | - | 1 | 460 | 40129539 |
Gene Symbol: Olfr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206822.1 | NP_996558.1 | 1 | 434 | ||
GenBank mRNA | BC148303.2 | - | 1 | 434 | 40041401 |
Gene Symbol: Olfr1393 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146471.1 | NP_666682.1 | 1 | 434 | ||
GenBank mRNA | BC104066.1 | - | 1 | 434 | 40041234 | |
BC104067.1 | - | 1 | 434 | 40041236 |

More Information
Other Species Targeted

Gene Ontology Categories:
