siRNA Details
Gene Symbol: Olfr959 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146508.1 | NP_666719.1 | 1 | 261 | ||
GenBank mRNA | BC120617.1 | - | 1 | 318 | 40129540 | |
BC120619.1 | - | 1 | 318 | 40129542 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
