siRNA Details
Gene Symbol: Olfr1360 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146543.2 | NP_666754.2 | 1 | 316 | ||
GenBank mRNA | BC130243.1 | - | 1 | 282 |
Gene Symbol: Olfr1359 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011820.1 | NP_001011820.1 | 1 | 278 | ||
GenBank mRNA | BC147114.1 | - | 1 | 307 | 9056158 | |
BC147115.1 | - | 1 | 306 | 9056125 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
