siRNA Details
Gene Symbol: Olfr800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146548.1 | NP_666759.1 | 1 | 261 | ||
GenBank mRNA | BC134373.1 | - | 1 | 265 | 40134856 |
Gene Symbol: Olfr777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146544.1 | NP_666755.1 | 1 | 261 | ||
GenBank mRNA | BC104276.1 | - | 1 | 261 | 40041253 | |
BC104277.1 | - | 1 | 256 |

More Information
Other Species Targeted

Gene Ontology Categories:
