siRNA Details
Gene Symbol: Olfr323 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146376.2 | NP_666488.2 | 1 | 824 | ||
GenBank mRNA | BC146611.1 | - | 2 | 264 |

More Information
Other Species Targeted

Gene Ontology Categories:
