siRNA Details
Gene Symbol: Olfr959 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146508.1 | NP_666719.1 | 1 | 467 | ||
GenBank mRNA | BC120617.1 | - | 1 | 524 | 40129540 | |
BC120619.1 | - | 1 | 524 | 40129542 |
Gene Symbol: Olfr934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146442.1 | NP_666653.1 | 1 | 467 | ||
GenBank mRNA | BC113124.1 | - | 1 | 466 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
