siRNA Details
Gene Symbol: Olfr704 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011749.1 | NP_001011749.1 | 1 | 341 |
Gene Symbol: Olfr695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146598.2 | NP_666809.1 | 3 | 594 | ||
GenBank mRNA | BC104092.1 | - | 1 | 341 | 40042972 | |
BC104093.1 | - | 1 | 341 | 40042973 | ||
BC129798.1 | - | 1 | 341 | 40042986 |

More Information
Other Species Targeted

Gene Ontology Categories:
