siRNA Details
Gene Symbol: Olfr323 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146376.2 | NP_666488.2 | 1 | 886 | ||
GenBank mRNA | BC146611.1 | - | 2 | 326 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001000012.
Gene Ontology Categories:
