siRNA Details
Gene Symbol: Olfr1392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146470.2 | NP_666681.1 | 1 | 940 | ||
GenBank mRNA | BC120614.1 | - | 1 | 936 | 40129537 | |
BC120616.1 | - | 1 | 940 | 40129539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
