siRNA Details
Gene Symbol: Olfr494 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146737.1 | NP_666948.1 | 1 | 88 | ||
GenBank mRNA | BC127987.1 | - | 92 | 40135591 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
