siRNA Details
Gene Symbol: Olfr935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146746.1 | NP_666957.1 | 1 | 268 |
Gene Symbol: Olfr146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146747.1 | NP_666958.1 | 1 | 268 | ||
GenBank mRNA | BC132188.1 | - | 1 | 348 | 40130465 | |
BC132190.1 | - | 1 | 348 | 40130467 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
