siRNA Details
Gene Symbol: PPIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000942.4 | NP_000933.1 | 1, 2 | 312 | ||
GenBank mRNA | AK291149.1 | - | 1, 2 | 176 | ||
BC001125.2 | - | 1, 2 | 161 | 2966791 | ||
BC008848.2 | - | 1, 2 | 283 | 3502055 | ||
BC020800.1 | - | 1, 2 | 168 | 4248828 | ||
BC032138.2 | - | 1, 2 | 166 | 4896656 | ||
BQ931949.1 | - | 1, 2 | 379 | |||
CR456829.1 | - | 1, 2 | 143 | |||
EU794651.1 | - | 1, 2 | 312 | |||
M60457.1 | - | 1, 2 | 117 | |||
M60857.1 | - | 1, 2 | 131 | |||
M63573.1 | - | 1, 2 | 164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
