siRNA Details
Gene Symbol: Olfr248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146714.2 | NP_666925.2 | 1 | 481 | ||
GenBank mRNA | BC131960.1 | - | 1 | 481 | 40130237 | |
BC131962.1 | - | 1 | 481 | 40130239 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
