siRNA Details
Gene Symbol: AK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005353.2 | NP_001005353.1 | 2, 3 | 501 | ||
NM_013410.3 | NP_037542.1 | 1, 2 | 384 | |||
NM_203464.2 | NP_982289.1 | 2, 3 | 390 | |||
GenBank mRNA | AK025926.1 | - | 1, 2 | 348 | ||
AK313611.1 | - | 1, 2 | 348 | |||
BC016180.1 | - | 1, 2 | 284 | 4637988 | ||
BC040224.1 | - | 1, 2 | 260 | 5188877 | ||
BC066944.1 | - | 2, 3 | 501 | 4819382 | ||
BC136886.1 | - | 1, 2 | 225 | 9020877 | ||
BC136887.1 | - | 1 | 225 | 9020878 | ||
BC146653.1 | - | 2, 3 | 375 | 40147793 | ||
BC148270.1 | - | 1, 2 | 165 | 8860213 | ||
BF205061.1 | - | 1 | 349 | |||
BG703687.1 | - | 2, 3 | 505 | |||
BM763432.1 | - | 2, 3 | 390 | |||
BU164554.1 | - | 2, 3 | 347 | |||
CR456830.1 | - | 1, 2 | 143 | |||
DB061266.1 | - | 1, 2 | 384 | |||
DR003868.1 | - | 2, 3 | 425 | |||
X60673.1 | - | 1, 2 | 326 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
