siRNA Details
Gene Symbol: Olfr1106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146752.2 | NP_666963.2 | 1 | 705 | ||
GenBank mRNA | BC127974.1 | - | 1 | 709 | 40135141 |

More Information
Other Species Targeted

Gene Ontology Categories:
