siRNA Details
Gene Symbol: Olfr1208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146778.1 | NP_666989.1 | 1 | 416 | ||
GenBank mRNA | BC150697.1 | - | 1 | 469 | 9087608 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
