siRNA Details
Gene Symbol: Olfr1444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146702.1 | NP_666913.1 | 1 | 281 | ||
GenBank mRNA | BC119059.1 | - | 1 | 316 | 8733808 | |
BC125619.1 | - | 1 | 316 | 40130134 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
