siRNA Details
Gene Symbol: Olfr482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146733.1 | NP_666944.1 | 1 | 806 |
Gene Symbol: Olfr506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011871.1 | NP_001011871.1 | 1 | 806 | ||
GenBank mRNA | BC147670.1 | - | 1 | 835 | 9056643 | |
BC147675.1 | - | 1 | 835 | 9056648 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
