siRNA Details
Gene Symbol: AK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005353.2 | NP_001005353.1 | 4 | 767 | ||
NM_013410.3 | NP_037542.1 | 3 | 650 | |||
NM_203464.2 | NP_982289.1 | 4 | 656 | |||
XM_017000612.1 | XP_016856101.1 | 3 | 402 | |||
XM_017000613.1 | XP_016856102.1 | 3 | 588 | |||
GenBank mRNA | AK025926.1 | - | 3 | 614 | ||
AK309057.1 | - | 3 | 358 | |||
AK313611.1 | - | 3 | 614 | |||
BC016180.1 | - | 3 | 550 | 4637988 | ||
BC040224.1 | - | 3 | 526 | 5188877 | ||
BC066944.1 | - | 4 | 767 | 4819382 | ||
BC136886.1 | - | 3 | 491 | 9020877 | ||
BC136887.1 | - | 1 | 491 | 9020878 | ||
BC146653.1 | - | 4 | 641 | 40147793 | ||
BC148270.1 | - | 3 | 431 | 8860213 | ||
BF205061.1 | - | 1 | 612 | |||
BU164554.1 | - | 4 | 613 | |||
CR456830.1 | - | 3 | 409 | |||
DR003868.1 | - | 4 | 689 | |||
X60673.1 | - | 3 | 592 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
