siRNA Details
Gene Symbol: Olfr1208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146778.1 | NP_666989.1 | 1 | 806 | ||
GenBank mRNA | BC150697.1 | - | 1 | 859 | 9087608 |
Gene Symbol: Olfr1198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207567.1 | NP_997450.1 | 1 | 806 |

More Information
Other Species Targeted

Gene Ontology Categories:
