siRNA Details
Gene Symbol: Olfr136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146807.1 | NP_667018.1 | 1 | 333 |
Gene Symbol: Olfr138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130868.1 | NP_570938.1 | 1 | 333 | ||
GenBank mRNA | BC104280.1 | - | 1 | 333 | 40041704 | |
BC104281.1 | - | 1 | 333 | 40041709 | ||
BC127129.1 | - | 1 | 333 | 40041712 | ||
BC127966.1 | - | 1 | 337 | 40134997 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
