siRNA Details
Gene Symbol: Olfr1161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146848.2 | NP_667059.2 | 1 | 741 | ||
GenBank mRNA | BC131999.1 | - | 1 | 741 | 40130276 | |
BC132001.1 | - | 1 | 741 | 40130278 |
