siRNA Details
Gene Symbol: Olfr900 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146874.1 | NP_667085.1 | 1 | 312 | ||
GenBank mRNA | BC106986.2 | - | 1 | 312 | 40043276 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
