siRNA Details
Gene Symbol: Olfr1298 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146886.1 | NP_667097.1 | 1 | 294 | ||
GenBank mRNA | BC104070.1 | - | 1 | 294 | ||
BC104071.1 | - | 1 | 294 | |||
BC119407.1 | - | 1 | 294 | |||
BC119408.1 | - | 1 | 293 | |||
BC126868.1 | - | 1 | 294 | 40044133 | ||
BC127217.1 | - | 1 | 294 | 40042024 | ||
BC127218.1 | - | 1 | 294 | 40042025 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
