siRNA Details
Gene Symbol: Olfr1297 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146888.1 | NP_667099.1 | 1 | 294 | ||
GenBank mRNA | BC104072.2 | - | 1 | 294 | 40041728 | |
BC104073.2 | - | 1 | 294 | 40041729 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
