siRNA Details
Gene Symbol: Olfr1426 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146809.2 | NP_667020.1 | 3 | 734 | ||
GenBank mRNA | BC058600.1 | - | 3 | 734 | 6390368 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
