siRNA Details
Gene Symbol: Olfr926 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146815.1 | NP_667026.1 | 1 | 416 | ||
GenBank mRNA | BC120558.1 | - | 1 | 468 | 40129481 | |
BC125490.1 | - | 1 | 468 | 40130005 |
Gene Symbol: Olfr924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207560.1 | NP_997443.1 | 1 | 416 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
