siRNA Details
Gene Symbol: Olfr1184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146823.1 | NP_667034.1 | 1 | 302 |
Gene Symbol: Olfr1181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011816.1 | NP_001011816.1 | 1 | 302 |
Gene Symbol: Olfr1182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011535.2 | NP_001011535.1 | 3 | 562 | ||
GenBank mRNA | DR065631.1 | - | 3 | 562 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
