siRNA Details
Gene Symbol: Olfr969 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146826.1 | NP_667037.1 | 1 | 227 |
Gene Symbol: Olfr970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146611.1 | NP_666822.1 | 1 | 227 |

More Information
Other Species Targeted

Gene Ontology Categories:
