siRNA Details
Gene Symbol: Olfr101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146834.1 | NP_667045.1 | 1 | 917 | ||
GenBank mRNA | BC138322.1 | - | 1 | 981 | 8861342 | |
BC138323.1 | - | 1 | 981 | 8861343 |
Gene Symbol: Olfr102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011721.2 | NP_001011721.2 | 1 | 972 | ||
GenBank mRNA | BC139114.1 | - | 1 | 972 | 8862136 | |
BC139115.1 | - | 1 | 972 | 8862137 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
