siRNA Details
Gene Symbol: Olfr1502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146797.1 | NP_667008.1 | 1 | 902 | ||
GenBank mRNA | BC106810.1 | - | 1 | 902 | 40042985 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
