siRNA Details
Gene Symbol: Olfr620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146812.2 | NP_667023.2 | 1 | 710 | ||
GenBank mRNA | BC106798.1 | - | 1 | 710 | 40042289 | |
BC106799.1 | - | 1 | 710 | 40042291 |
Gene Symbol: Olfr624 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011865.2 | NP_001011865.2 | 1 | 692 |

More Information
Other Species Targeted

Gene Ontology Categories:
