siRNA Details
Gene Symbol: Olfr330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146879.2 | NP_667090.1 | 2 | 943 | ||
GenBank mRNA | BC120838.1 | - | 1 | 1003 | 40129761 | |
BC172163.1 | - | 1 | 921 | |||
BC172164.1 | - | 1 | 921 | |||
BC172166.1 | - | 1 | 921 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
