siRNA Details
Gene Symbol: Olfr790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146933.1 | NP_667144.1 | 1 | 288 | ||
GenBank mRNA | BC125495.1 | - | 1 | 361 | 40130010 | |
BC132182.1 | - | 1 | 361 | 40130459 |

More Information
Other Species Targeted

Gene Ontology Categories:
