siRNA Details
Gene Symbol: Olfr347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146943.1 | NP_667154.1 | 1 | 893 | ||
GenBank mRNA | BC127977.1 | - | 1 | 897 | 40135187 |
Gene Symbol: Olfr344 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146628.1 | NP_666839.1 | 1 | 893 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
