siRNA Details
Gene Symbol: Olfr531 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146953.1 | NP_667164.1 | 1 | 120 | ||
GenBank mRNA | BC104052.1 | - | 1 | 120 | 40040241 | |
BC104053.1 | - | 1 | 120 | 40040242 |

More Information
Other Species Targeted

Gene Ontology Categories:
