siRNA Details
Gene Symbol: Olfr1226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146967.1 | NP_667178.1 | 1 | 338 | ||
GenBank mRNA | BC106781.1 | - | 1 | 338 | 40040933 | |
BC106782.1 | - | 1 | 338 | 40040934 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
