siRNA Details
Gene Symbol: Olfr38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146986.1 | NP_667197.1 | 1 | 400 | ||
GenBank mRNA | BC106817.1 | - | 1 | 399 | ||
BC128054.1 | - | 1 | 400 | 40043588 |
Gene Symbol: Olfr452 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011869.1 | NP_001011869.1 | 1 | 400 |

More Information
Other Species Targeted

Gene Ontology Categories:
