siRNA Details
Gene Symbol: Olfr447 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146988.1 | NP_667199.1 | 1 | 883 | ||
GenBank mRNA | BC104271.1 | - | 1 | 883 | 40040950 | |
BC104272.1 | - | 1 | 881 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
