siRNA Details
Gene Symbol: Olfr845 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207145.2 | NP_997028.1 | 1 | 712 | ||
GenBank mRNA | BC127982.1 | - | 1 | 713 | 40135240 |
Gene Symbol: Olfr851 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146905.2 | NP_667116.2 | 1 | 709 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
