siRNA Details
Gene Symbol: Olfr32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010980.2 | NP_035110.1 | 5 | 1161 | ||
GenBank mRNA | BC119404.1 | - | 1 | 936 | 8734153 | |
BC120653.1 | - | 1 | 936 | 40129576 |
Gene Symbol: Olfr1270 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146985.2 | NP_667196.1 | 1 | 913 | ||
GenBank mRNA | BC119113.1 | - | 1 | 913 | 8733862 | |
BC119115.1 | - | 1 | 913 | 8733864 |

More Information
Other Species Targeted

Gene Ontology Categories:
