siRNA Details
Gene Symbol: Olfr38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146986.1 | NP_667197.1 | 1 | 436 | ||
GenBank mRNA | BC106817.1 | - | 1 | 435 | ||
BC128054.1 | - | 1 | 436 | 40043588 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
