siRNA Details
Gene Symbol: Olfr206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146991.1 | NP_667202.1 | 1 | 890 | ||
GenBank mRNA | BC138396.1 | - | 1 | 917 | 8861416 | |
BC138397.1 | - | 1 | 917 | 8861417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
