siRNA Details
Gene Symbol: RRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033.4 | NP_001024.1 | 3 | 575 | ||
GenBank mRNA | AK222786.1 | - | 3 | 455 | ||
AK297988.1 | - | 3 | 320 | |||
BC006498.2 | - | 3 | 406 | 2989344 | ||
BP229960.1 | - | 3 | 566 | |||
X59543.1 | - | 3 | 434 | |||
X59617.1 | - | 3 | 428 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
