siRNA Details
Gene Symbol: Olfr1417 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146936.1 | NP_667147.1 | 1 | 827 | ||
GenBank mRNA | BC148189.1 | - | 1 | 827 | 40051854 | |
BC153816.1 | - | 1 | 827 | 40043087 |
Gene Symbol: Olfr1418 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011524.1 | NP_001011524.1 | 1 | 827 | ||
GenBank mRNA | BC150977.1 | - | 1 | 877 | 9087890 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
