siRNA Details
Gene Symbol: Olfr531 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146953.1 | NP_667164.1 | 1 | 881 | ||
GenBank mRNA | BC104052.1 | - | 1 | 881 | 40040241 | |
BC104053.1 | - | 1 | 881 | 40040242 |
Gene Symbol: Olfr527 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011776.1 | NP_001011776.1 | 1 | 884 | ||
GenBank mRNA | BC147213.1 | - | 1 | 905 | 9056261 | |
BC147214.1 | - | 1 | 905 | 9056246 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
