siRNA Details
Gene Symbol: Olfr1228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146971.1 | NP_667182.1 | 1 | 495 | ||
GenBank mRNA | BC125451.1 | - | 1 | 578 | 40129966 | |
BC125453.1 | - | 1 | 578 | 40129968 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
